Alfa Aesar™ Potassium iodate, Acculute Standard Volumetric Solution, Final Concentration 0.1N

Manufacturer: Alfa Aesar™ 88605LS

Quantity 6 units

 View more versions of this product

Catalog No. AA88605LS

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7758-05-6
    Molecular Formula KIO3
    Formula Weight 214
    MDL Number MFCD00011406
    UN Number UN3139
    Chemical Name or Material Potassium iodate Acculute Standard Volumetric Solution, Final Concentration 0.1N

    Specifications

    Formula Weight 214
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 6 units
    Physical Form Liquid
    CAS 7758-05-6
    Health Hazard 2 GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    TSCA TSCA
    MDL Number MFCD00011406
    UN Number UN3139
    Health Hazard 1 Warning
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING LIQUID, N.O.S.
    Molecular Formula KIO3
    Odor Odorless
    Chemical Name or Material Potassium iodate Acculute Standard Volumetric Solution, Final Concentration 0.1N

    GHS H Statement
    H272-H315-H319-H335
    May intensify fire
    oxidizer.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P221-P210-P220-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Keep/Store away from clothing/combustible materials.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit