Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
2.5kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle

Chemical Identifiers

7758-05-6
IKO3
214.00
JLKDVMWYMMLWTI-UHFFFAOYSA-M
23665710

High quality chemicals for laboratory and specialized industrial use

SMILES [O-]I(=O)=O.[K+]
IUPAC Name potassium;iodate
Formula Weight 214
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 3.89kg
Quantity 500g
Chemical Name or Material Potassium Iodate
Grade Baker Analyzed™ ACS Reagent