Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodate, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
2.5kg
500g
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle

Chemical Identifiers

7758-05-6
IKO3
214.00
JLKDVMWYMMLWTI-UHFFFAOYSA-M
23665710

High quality chemicals for laboratory and specialized industrial use

CAS 7758-05-6
Molecular Formula IKO3
InChI Key JLKDVMWYMMLWTI-UHFFFAOYSA-M
SMILES [O-]I(=O)=O.[K+]
IUPAC Name potassium;iodate
Molecular Weight (g/mol) 214.00
PubChem CID 23665710
Formula Weight 214
Packaging Poly Bottle
Specific Gravity 1L = 3.89kg
Quantity 125g
Chemical Name or Material Potassium Iodate
Grade Baker Analyzed™ ACS Reagent
Show More Show Less