Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodate, FCC, 99-101%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7758-05-6
IKO3
214.00
JLKDVMWYMMLWTI-UHFFFAOYSA-M
potassium iodate
[K+].[O-][I](=O)=O
7758-05-6
IKO3
214.00
JLKDVMWYMMLWTI-UHFFFAOYSA-M
potassium iodate
[K+].[O-][I](=O)=O

P1309, 7758-05-6, KIO3

Spectrum™ Potassium Iodate, FCC is used as a source for dietary iodine. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade FCC
CAS Max % 1
Percent Purity 99 to 101%
Chlorate To pass test
Iodide To pass test
Lead (Pb) 4 mg/kg
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 25 g