Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodate-Iodide, 0.0125 Normal, 1mL = 0.5mg SO32-, Ricca Chemical

Click to view available options
Quantity:
1L
Packaging:
Glass amber
7758-05-6,7681-11-0,7758-05-6,144-55-8
IKO3
214.00
JLKDVMWYMMLWTI-UHFFFAOYSA-M
potassium iodate,potassium triodate,caswell no. 693a,iodic acid, potassium salt,unii-i139e44nhl,potassium iodine oxide kio3,iodic acid hio3 , potassium salt,epa pesticide chemical code 075703,iodic acid hio3 , potassium salt 1:1,kaliumjodat
23665710
potassium;iodate
[O-]I(=O)=O.[K+]
7758-05-6,7681-11-0,7758-05-6,144-55-8
IKO3
214.00
JLKDVMWYMMLWTI-UHFFFAOYSA-M
potassium iodate,potassium triodate,caswell no. 693a,iodic acid, potassium salt,unii-i139e44nhl,potassium iodine oxide kio3,iodic acid hio3 , potassium salt,epa pesticide chemical code 075703,iodic acid hio3 , potassium salt 1:1,kaliumjodat
23665710
potassium;iodate
[O-]I(=O)=O.[K+]
CAS Min % 98.0,0.42,0.04,0.03
CAS Max % 100.0,0.44,0.05,0.03
Concentration or Composition (by Analyte or Components) 0.0125 Normal, 1mL = 0.5mg SO32-
Physical Form Liquid
Packaging Glass amber
Color Colorless
Quantity 1L
Chemical Name or Material Potassium Iodate-Iodide

ShelfLife : 12 months