Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium iodide, 1:2 w/w soln. in distilled water

Manufacturer:  Alfa Aesar™ 47293K2

Quantity 1L

 View more versions of this product

Catalog No. AA47293K2


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7681-11-0
MFCD00011405
Potassium iodide
166.01
14,7643

Specifications

7681-11-0
166.01
14,7643
1.677
1330°C
681°C
1L
Potassium iodide
Liquid
MFCD00011405
Odorless
3.12g/mL
Colorless
1:2 w/w soln. in distilled water
Hygroscopic

Safety and Handling

SDS & Safety Documents

RTECSNumber : TT2975000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit