Potassium Iodide, 2% (w/v) Aqueous Solution, Ricca Chemical

Manufacturer: Ricca Chemical CompanySupplier Diversity Partner 62885

Packaging Cubitainer
Quantity 20L

 View more versions of this product

Catalog No. 62885

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7732-18-5,7681-11-0,144-55-8
    CAS Min % 96.0,1.92,0.01
    CAS Max % 100.0,2.0,0.01
    Physical Form Liquid
    Chemical Name or Material Potassium Iodide

    Specifications

    Color Clear colorless
    Physical Form Liquid
    Quantity 20L
    Concentration or Composition Notes 2% (w/v)
    Solvent or Matrix Aqueous Solution
    CAS Max % 0.01,100.0,2.0
    CAS 7732-18-5,7681-11-0,144-55-8
    CAS Min % 96.0,1.92,0.01
    Packaging Cubitainer
    Chemical Name or Material Potassium Iodide

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit