Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium iodide, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
7681-11-0
IK
166
MFCD00011405
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium iodide,potassium iodide ki,pima,kali iodide,thyro-block,knollide,kisol,asmofug e,potassium diiodide,potassium monoiodide
4875
CHEBI:8346
potassium;iodide
[K+].[I-]
7681-11-0
IK
166
MFCD00011405
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium iodide,potassium iodide ki,pima,kali iodide,thyro-block,knollide,kisol,asmofug e,potassium diiodide,potassium monoiodide
4875
CHEBI:8346
potassium;iodide
[K+].[I-]
Alkalinity 0.022 % max. (as KOH)
Assay Percent Range 99% min. (on dry substance) (Argentometry)
Linear Formula KI
Fieser 06,227; 11,440
Merck Index 15, 7764
Solubility Information Solubility in water: 1.43 kg/l. Other solubilities: 45g/L ethanol
Formula Weight 166
Physical Form Flakes or Powder
Percent Purity 99+%
Grade Analysis
Density 3.13
Heavy Metals (as Pb) 10ppm max.
Iodate 2ppm max.
Iron (Fe) 5ppm max.
Loss on Drying 0.2 % max. (150°C, 6 hrs)
Nitrogen Compounds 10ppm (as N) max.
Packaging Glass bottle
Solubility Solubility in water: 1.43 kg/l. Other solubilities: 45g/L ethanol
Sulfate 50ppm max.
Boiling Point 1330.0°C
Color Tan to White
Melting Point 680.0°C
pH 6.0 to 9.2 (5% soln. at 25°C)
Quantity 2.5kg
Chemical Name or Material Potassium iodide
Show More Show Less

GHS Signal Word: Danger

EINECSNumber : 231-659-4