Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium iodide, 99%, extra pure, briquettes, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
10kg
Packaging:
Glass bottle
Plastic drum
7681-11-0
IK
166
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium iodide,potassium iodide ki,pima,kali iodide,thyro-block,knollide,kisol,asmofug e,potassium diiodide,potassium monoiodide
4875
CHEBI:8346
potassium;iodide
[K+].[I-]
7681-11-0
IK
166
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium iodide,potassium iodide ki,pima,kali iodide,thyro-block,knollide,kisol,asmofug e,potassium diiodide,potassium monoiodide
4875
CHEBI:8346
potassium;iodide
[K+].[I-]
Assay Percent Range 98.5% min. (on dried substance) (Argentometry)
Linear Formula KI
Fieser 06,227; 11,44
Merck Index 15,7764
Solubility Information Solubility in water: 1.43 kg/l. Other solubilities: 45g/L ethanol
Formula Weight 166
Physical Form Briquettes
Percent Purity 99%
Grade Extra Pure
Heavy Metals (as Pb) 10ppm max.
Iodate 4ppm max.
Iron (Fe) 20ppm max.
Loss on Drying 1% max.
Packaging Glass bottle
Solubility Solubility in water: 1.43 kg/l. Other solubilities: 45g/L ethanol
Sulfate 200ppm max.
Thiosulphate 100ppm max.
Boiling Point 1330.0°C
Color White
Melting Point 680.0°C
Quantity 2.5kg
Chemical Name or Material Potassium iodide
Show More Show Less

GHS Signal Word: Danger

EINECSNumber : 231-659-4