Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

7681-11-0
IK
166.003
NLKNQRATVPKPDG-UHFFFAOYSA-M
4875
CHEBI:8346

High quality chemicals for laboratory and specialized industrial use

CAS 7681-11-0
Molecular Formula IK
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES [K+].[I-]
IUPAC Name potassium;iodide
Molecular Weight (g/mol) 166.003
PubChem CID 4875
ChEBI CHEBI:8346
Formula Weight 166
Physical Form Granular
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 3.10kg
Quantity 500g
Chemical Name or Material Potassium Iodide
Grade Baker Analyzed™ ACS Reagent
Show More Show Less