Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, Granular, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Wide Mouth Amber Glass Bottle
7681-11-0
IK
166.003
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium iodide,potassium iodide ki,pima,kali iodide,thyro-block,knollide,kisol,asmofug e,potassium diiodide,potassium monoiodide
4875
CHEBI:8346
potassium;iodide
[K+].[I-]
7681-11-0
IK
166.003
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium iodide,potassium iodide ki,pima,kali iodide,thyro-block,knollide,kisol,asmofug e,potassium diiodide,potassium monoiodide
4875
CHEBI:8346
potassium;iodide
[K+].[I-]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 166
Physical Form Granular
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 3.10kg
Quantity 2.5kg
Chemical Name or Material Potassium Iodide