Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Iodide, Granular, Reagent, ACS, 99%, Spectrum™

Click to view available options
Quantity:
25 g
125 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
7681-11-0
IK
166.00
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium iodide
[K+].[I-]
7681-11-0
IK
166.00
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium iodide
[K+].[I-]

P1340, 7681-11-0, KI

Spectrum™ Potassium Iodide, Granular, Reagent, ACS is refered to by the chemical formula KI and is an inorganic compound. Used as a component in film photography before the discovery of silver iodide, it is also used in the electrolyte of dye sensitized solar cells. KI is also used as an emergency blockade of radio-iodine, a common form of radiation poisoning. As an ACS grade Reagent, Spectrum Chemical manufactured potassium iodide is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Grade ACS
CAS Max % 1
Barium (Ba) 2e-005
Percent Purity 99%
Calcium (Ca) 2e-005
Heavy Metals (as Pb) 5 ppm
Insoluble Matter 5e-005
Iron (Fe) 3 ppm
Loss on Drying 0.002
Magnesium (Mg) 1e-005
Packaging Poly Pail
Sodium (Na) 5e-005
Sulfate 5e-005
pH 6.0 to 9.2
Quantity 12 kg
Show More Show Less