Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium meta-Periodate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
125g
500g
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle

Chemical Identifiers

7790-21-8
IKO4
229.999
FJVZDOGVDJCCCR-UHFFFAOYSA-M
516896

High quality chemicals for laboratory and specialized industrial use

SMILES [O-]I(=O)(=O)=O.[K+]
IUPAC Name potassium;periodate
Formula Weight 230
Physical Form Crystals
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 3.62kg
Quantity 500g
Chemical Name or Material Potassium meta-Periodate
Grade Baker Analyzed™ ACS Reagent