Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Metabisulfite, Crystal, NF, 51.8-57.6%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
16731-55-8
K2O5S2
222.31
RWPGFSMJFRPDDP-UHFFFAOYSA-L
dipotassium sulfinatosulfonate
[K+].[K+].[O-]S(=O)S([O-])(=O)=O
16731-55-8
K2O5S2
222.31
RWPGFSMJFRPDDP-UHFFFAOYSA-L
dipotassium sulfinatosulfonate
[K+].[K+].[O-]S(=O)S([O-])(=O)=O

P1346, 16731-55-8, K2S2O5

Spectrum™ Potassium Metabisulfite, Crystal, NF is used primarily as an antioxidant in pharmaceutical formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Pail
Quantity 12 kg
CAS Max % 1
Percent Purity 51.8 to 57.6%
Grade NF