Alfa Aesar™ Potassium methoxide, 5% w/v in methanol

Manufacturer: Alfa Aesar™ 42551AE

Quantity 100mL

 View more versions of this product

Catalog No. AA42551AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 865-33-8
    Linear Formula KOCH3
    Formula Weight 70.14
    MDL Number MFCD00012416
    UN Number UN3274
    Chemical Name or Material Potassium methoxide

    Specifications

    Formula Weight 70.14
    Health Hazard 2 GHS H Statement
    H225-H301-H311-H331-H370-H314-H318
    Highly flammable liquid and vapor.
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes damage to organs.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Physical Form Liquid
    MDL Number MFCD00012416
    Health Hazard 1 Danger
    Concentration 5% w/v in methanol
    Quantity 100mL
    UN Number UN3274
    Sensitivity Air & Moisture Sensitive
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.
    TSCA TSCA
    Linear Formula KOCH3
    CAS 865-33-8
    Odor Alcohol-like
    Chemical Name or Material Potassium methoxide

    GHS H Statement
    H225-H301-H311-H331-H370-H314-H318
    Highly flammable liquid and vapor.
    Toxic if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes damage to organs.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P280-P303+P361+P353-P305+P351+P338-P310
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit