Alfa Aesar™ Potassium methoxymethyltrifluoroborate, 95%

Manufacturer: Alfa Aesar™ H3267404

Quantity 2g

 View more versions of this product

Catalog No. AAH3267404

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 910251-11-5
    Molecular Formula C2H5BF3KO
    Formula Weight 151.97
    MDL Number MFCD10566517
    Chemical Name or Material Potassium methoxymethyltrifluoroborate

    Specifications

    Formula Weight 151.97
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C2H5BF3KO
    MDL Number MFCD10566517
    CAS 910251-11-5
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 2g
    Chemical Name or Material Potassium methoxymethyltrifluoroborate
    Assay 95%

    Description

    Applications
    Potassium methoxymethyltrifluoroborate is used in the cross-coupling of mesylated phenol derivatives. The corresponding aryl- and heteroaryl alkoxymethyl compounds are prepared with both electron-rich and electron-poor substituents on the activated alcohol.

    Notes
    Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit