Potassium molybdenum oxide, anhydrous, 99.8% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 2289809

Quantity 10g

 View more versions of this product

Catalog No. AA2289809

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13446-49-6
    Physical Form Powder
    Assay Percent Range 99.8% (metals basis)
    Molecular Formula K2MoO4
    Formula Weight 238.14
    MDL Number MFCD00011370
    Chemical Name or Material Potassium molybdenum oxide, anhydrous

    Specifications

    Physical Form Powder
    Formula Weight 238.14
    Quantity 10g
    Color White
    EINECS Number 236-599-2
    Anhydrous or Hydrate Anhydrous
    Odor Odorless
    Assay Percent Range 99.8% (metals basis)
    Molecular Formula K2MoO4
    Appearance White powder
    MDL Number MFCD00011370
    CAS 13446-49-6
    Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Health Hazard 2 In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
    Chemical Name or Material Potassium molybdenum oxide, anhydrous

    In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.

    H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit