Alfa Aesar™ Potassium niobium oxide, Puratronic™, 99.999% (metals basis)

Manufacturer: Alfa Aesar™ 1084314

Quantity 25g

 View more versions of this product

Catalog No. AA1084314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12030-85-2
    Formula Weight 180.01
    MDL Number MFCD00049651
    Synonym Potassium niobate
    Chemical Name or Material Potassium niobium oxide
    Grade Puratronic™

    Specifications

    Formula Weight 180.01
    Physical Form Powder
    MDL Number MFCD00049651
    Synonym Potassium niobate
    TSCA TSCA
    CAS 12030-85-2
    Quantity 25g
    RTECS Number TT3500000
    Chemical Name or Material Potassium niobium oxide
    Assay 99.999%, (metals basis)
    Grade Puratronic™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit