Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Fluka™ Potassium Nitrate, ≥99.0%, ACS Reagent, Honeywell™ Fluka™

Click to view available options
Quantity:
100g
500g
2.5kg
12kg
Packaging:
Drum with inliner
Poly Bottle
7757-79-1
KNO3
101.102
MFCD00011409
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium nitrate,saltpeter,nitre,nitrate of potash,vicknite,nitric acid potassium salt,nitric acid, potassium salt,saltpetre,kaliumnitrat,kalii nitras
24434
CHEBI:63043
potassium;nitrate
[N+](=O)([O-])[O-].[K+]
7757-79-1
KNO3
101.102
MFCD00011409
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium nitrate,saltpeter,nitre,nitrate of potash,vicknite,nitric acid potassium salt,nitric acid, potassium salt,saltpetre,kaliumnitrat,kalii nitras
24434
CHEBI:63043
potassium;nitrate
[N+](=O)([O-])[O-].[K+]

ACS Reagent,≥99.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
UN Number UN1486
Formula Weight 101.1g/mol
Percent Purity ≥99.0%
Grade ACS Reagent
Packaging Poly Bottle
Quantity 100g
Chemical Name or Material Potassium Nitrate

P220-P273

H272-H412

DOTInformation : 5.1/PGIII

EINECSNumber : 231-818-8

RTECSNumber : TT3700000