Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium nitrate, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
500g
1kg
2.5kg
10kg
Packaging:
Plastic Drum
Plastic bottle
7757-79-1
KNO3
101.1
MFCD00011409
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium nitrate,saltpeter,nitre,nitrate of potash,vicknite,nitric acid potassium salt,nitric acid, potassium salt,saltpetre,kaliumnitrat,kalii nitras
24434
CHEBI:63043
potassium;nitrate
[N+](=O)([O-])[O-].[K+]
7757-79-1
KNO3
101.1
MFCD00011409
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium nitrate,saltpeter,nitre,nitrate of potash,vicknite,nitric acid potassium salt,nitric acid, potassium salt,saltpetre,kaliumnitrat,kalii nitras
24434
CHEBI:63043
potassium;nitrate
[N+](=O)([O-])[O-].[K+]
Assay Percent Range 99+%
Linear Formula KNO3
Fieser 05,561
Merck Index 15,7769
Solubility Information Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol
Formula Weight 101.1
Physical Form Fine Crystals
Percent Purity 99+%
Grade Analysis
Chloride 20ppm max
Chlorine (Cl) 20ppm max.
Heavy Metals (as Pb) 10ppm max.
Iodate 5ppm max.
Iron (Fe) 10ppm max.
Nitrite 10ppm max.
Packaging Plastic Drum
Sodium (Na) 200ppm max.
Solubility Solubility in water: 320g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol
Sulfate 50ppm max.
Boiling Point 400.0°C
Color White
Melting Point 334.0°C
pH 5.5 to 8 (5% soln. at 20°C)
Quantity 10kg
Chemical Name or Material Potassium nitrate
Show More Show Less

GHS H Statement:
May intensify fire; oxidizer.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 231-818-8