Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Nitrate, BiotechGrade, 99.5%, Spectrum™

Click to view available options
Quantity:
500 g
6 x 500 g
Packaging:
Amber Glass Bottle
7757-79-1
KNO3
101.10
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium nitrate
[K+].[O-][N+]([O-])=O
7757-79-1
KNO3
101.10
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium nitrate
[K+].[O-][N+]([O-])=O

P1352, 7757-79-1, KNO3

Spectrum™ Potassium Nitrate, BiotechGrade - this compound has been tested to alleviate NaCl stress symptoms in plants. Mechanistic studies suggest that this may be achieved through reduction of electrolyte leakage, malondialdehyde and soluble sugar contents and improving the activities of antioxidant enzymes. Spectrum offers highly pure reagents suitable for biochemical research and analysis. The critical parameters involved are absence of inhibitors such as traces of heavy metals as well as biochemical function tests for enzymes, coenzymes and enzyme substrates.

CAS Max % 1
Percent Purity 99.5%
Cadmium (Cd) 5e-006
Calcium (Ca) 5e-005
Chromium (Cr) 5e-006
Copper (Cu) 5e-006
Insoluble Matter 0.0001
Loss on Drying 0.001
Magnesium (Mg) 5e-005
Manganese (Mn) 5e-006
Packaging Amber Glass Bottle
Sodium (Na) 5e-005
Sulfate 2e-005
Zinc (Zn) 5e-006
pH 5.0 to 8.0
Quantity 6 x 500 g
Show More Show Less