Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Nitrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle

Chemical Identifiers

7757-79-1
KNO3
101.102
FGIUAXJPYTZDNR-UHFFFAOYSA-N
24434
CHEBI:63043

High quality chemicals for laboratory and specialized industrial use

CAS 7757-79-1
Molecular Formula KNO3
InChI Key FGIUAXJPYTZDNR-UHFFFAOYSA-N
SMILES [N+](=O)([O-])[O-].[K+]
IUPAC Name potassium;nitrate
Molecular Weight (g/mol) 101.102
PubChem CID 24434
ChEBI CHEBI:63043
Formula Weight 101.09
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 2.10kg
Quantity 2.5kg
Chemical Name or Material Potassium Nitrate
Grade Baker Analyzed™ ACS Reagent
Show More Show Less