Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Nitrate, Crystal, FCC, 99-100.5%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
7757-79-1
KNO3
101.10
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium nitrate
[K+].[O-][N+]([O-])=O
7757-79-1
KNO3
101.10
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium nitrate
[K+].[O-][N+]([O-])=O

PO193, 7757-79-1, KNO3

Spectrum™ Potassium Nitrate, Crystal, FCC , also called salt peter, has been used as a food preservative and in toothpaste formulations for sensitive teeth. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage, nutritional supplement and personal care applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Grade FCC
CAS Max % 1
Percent Purity 99.0 to 100.5%
Chlorate To pass test
Lead (Pb) 4 mg/kg
Loss on Drying 0.01
Packaging Poly Pail
Quantity 12 kg