Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium osmate(VI) dihydrate, 51.0-52.0% Os, ACROS Organics™

Click to view available options
Quantity:
500mg
2.5g
Packaging:
Glass bottle
10022-66-9
K2O4Os·2H2O
368.42
MFCD00149919
DGODWNOPHMXOTR-UHFFFAOYSA-N
potassium osmate vi dihydrate,potassium dioxidodioxoosmium dihydrate,potassium osmate dihydrate,unii-5m2bwq2tyj,5m2bwq2tyj,osmate oso42-, dipotassium, dihydrate, t-4,dipotassium tetrahydroxodioxoosmate,potassium osmate k2oso2 oh 4,potassium osmate k2oso4 , dihydrate
53393272
dipotassium;dioxido(dioxo)osmium;dihydrate
O.O.[O-][Os](=O)(=O)[O-].[K+].[K+]
10022-66-9
K2O4Os·2H2O
368.42
MFCD00149919
DGODWNOPHMXOTR-UHFFFAOYSA-N
potassium osmate vi dihydrate,potassium dioxidodioxoosmium dihydrate,potassium osmate dihydrate,unii-5m2bwq2tyj,5m2bwq2tyj,osmate oso42-, dipotassium, dihydrate, t-4,dipotassium tetrahydroxodioxoosmate,potassium osmate k2oso2 oh 4,potassium osmate k2oso4 , dihydrate
53393272
dipotassium;dioxido(dioxo)osmium;dihydrate
O.O.[O-][Os](=O)(=O)[O-].[K+].[K+]
Linear Formula K2OsO4·2H2O
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol and ether
Formula Weight 368.42
Physical Form Crystalline Powder
Percent Purity 51.0-52% Os
Packaging Glass bottle
Solubility Solubility in water: soluble. Other solubilities: insoluble in alcohol and ether
Color Violet
Quantity 500mg
Chemical Name or Material Potassium osmate(VI) dihydrate, 51.0-52.0% Os

GHS H Statement
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
May intensify fire; oxidizer.
May be corrosive to metals.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Take any precaution to avoid mixing with combustibles.
In case of major

GHS Signal Word: Danger

EINECSNumber : 243-247-1

TSCA : TSCA