Learn More
Potassium oxalate hydrate, 99%, Thermo Scientific Chemicals
Description
Potassium oxalate hydrate is used as a chelating agent, a purifying agent in the pharmaceutical industry and a precipitating agent in rare-earth metal processing. It finds application as a grinding agent, as a rust remover in metal treatment and as a bleaching agent in the textile and wood industry. It is also used in waste water treatment and as a scale remover in automobile radiators. Furthermore, it is used as a reagent in analytical chemistry for the determination of calcium and other heavy metals as well as reducing agents in photography. It is also involved in the preparation of SBA-15 (nanostructured SiO2) supported vanadium oxide catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium oxalate hydrate is used as a chelating agent, a purifying agent in the pharmaceutical industry and a precipitating agent in rare-earth metal processing. It finds application as a grinding agent, as a rust remover in metal treatment and as a bleaching agent in the textile and wood industry. It is also used in waste water treatment and as a scale remover in automobile radiators. Furthermore, it is used as a reagent in analytical chemistry for the determination of calcium and other heavy metals as well as reducing agents in photography. It is also involved in the preparation of SBA-15 (nanostructured SiO2) supported vanadium oxide catalyst.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with halogens, ammonia, cyanides and heavy metals.
Specifications
Melting Point | >300°C |
Density | 2.13 |
Assay Percent Range | 99% |
Quantity | 1000 g |
Beilstein | 3752576 |
Sensitivity | Hygroscopic |
Merck Index | 14,7651 |
Solubility Information | Soluble in water. |
Formula Weight | 166.22 (Anhydrous) |
Percent Purity | 99% |
Show More |
Safety and Handling
Hazard Category | H302+H312 |
Hazard Statement | GHS H Statement H302-H312 |
Precautionary Statement | P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c |
EINECSNumber | 209-506-8 |
RTECSNumber | RO2885000 |
TSCA | Yes |
storageNote1 | Hygroscopic |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.