Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Oxalate, Monohydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle

Chemical Identifiers

6487-48-5
C2H2K2O5
184.23
QCPTVXCMROGZOL-UHFFFAOYSA-L
2724193

High quality chemicals for laboratory and specialized industrial use

SMILES C(=O)(C(=O)[O-])[O-].O.[K+].[K+]
IUPAC Name dipotassium;oxalate;hydrate
Formula Weight 184.23
Physical Form Crystals
Packaging Poly Bottle
Specific Gravity 1L = 2.13kg
Quantity 500g
Chemical Name or Material Potassium Oxalate, Monohydrate
Grade Baker Analyzed™ ACS Reagent