Alfa Aesar™ Potassium oxide, super, 96.5%

Manufacturer: Alfa Aesar™ 1405830

Quantity 250g

 View more versions of this product

Catalog No. AA1405830

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12030-88-5
    Formula Weight 71.1
    MDL Number MFCD00082411
    UN Number UN2466
    Synonym Potassium superoxide
    Chemical Name or Material Potassium oxide, Super

    Specifications

    Formula Weight 71.1
    Health Hazard 3 GHS P Statement
    P221-P283-P303+P361+P353-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Wear fire/flame resistant/retardant clothing.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Powder
    CAS 12030-88-5
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H271-H314-H318
    May cause fire or explosion
    strong oxidizer.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Density 2.14g/mL
    Odor Odorless
    MDL Number MFCD00082411
    Quantity 250g
    Melting Point ≈400°C
    DOT Information Transport Hazard Class: 5.1; Packing Group: I; Proper Shipping Name: POTASSIUM SUPEROXIDE
    TSCA TSCA
    Particle Size 2 to 10 Micron
    Sensitivity Air & Moisture Sensitive
    UN Number UN2466
    Synonym Potassium superoxide
    Chemical Name or Material Potassium oxide, Super
    Assay 96.5%

    GHS H Statement
    H271-H314-H318
    May cause fire or explosion
    strong oxidizer.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P221-P283-P303+P361+P353-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Wear fire/flame resistant/retardant clothing.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit