Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium pentaborate octahydrate, 97%

Manufacturer:  Alfa Aesar™ 39437A3

Quantity 2kg

 View more versions of this product

Catalog No. AA39437A3


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

12229-13-9
MFCD00213879
Potassium pentaborate octahydrate
586.43 (442.30 Anhydrous)
Potassium borate

Specifications

12229-13-9
586.43 (442.30 Anhydrous)
Potassium borate
White
2kg
97%
Solid
MFCD00213879
Insoluble in water
780°C
Potassium pentaborate octahydrate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H360
May damage fertility or the unborn child.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit