Alfa Aesar™ Potassium pentaborate octahydrate, 97%

Manufacturer: Alfa Aesar™ 3943736

Quantity 500g

 View more versions of this product

Catalog No. AA3943736

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12229-13-9
    Formula Weight 586.43 (442.30 Anhydrous)
    MDL Number MFCD00213879
    Synonym Potassium borate
    Chemical Name or Material Potassium pentaborate octahydrate

    Specifications

    Formula Weight 586.43 (442.30 Anhydrous)
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Solid
    MDL Number MFCD00213879
    Color White
    Melting Point 780°C
    Quantity 500g
    Health Hazard 1 Danger
    TSCA TSCA
    CAS 12229-13-9
    Synonym Potassium borate
    Solubility Information Insoluble in water
    Health Hazard 2 GHS H Statement
    H360
    May damage fertility or the unborn child.
    Chemical Name or Material Potassium pentaborate octahydrate
    Assay 97%

    GHS H Statement
    H360
    May damage fertility or the unborn child.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit