Alfa Aesar™ Potassium pentachlororhodate(III), Premion™, 99.99% (metals basis), Rh 28.2% min

Manufacturer: Alfa Aesar™ 4040906

Quantity 5g

 View more versions of this product

Catalog No. AA4040906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 65980-75-8
    Formula Weight 358.37
    MDL Number MFCD00049661
    Chemical Name or Material Potassium pentachlororhodate(III)
    Grade Premion™

    Specifications

    Formula Weight 358.37
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P301+P312-P321-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Specific treatment (see ... on this label).Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 65980-75-8
    Solubility Information Slightly soluble in cold water; Insoluble in alcohol
    Health Hazard 2 GHS H Statement
    H302-H315-H319
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    RTECS Number VI8580332
    Physical Form Powder
    MDL Number MFCD00049661
    Health Hazard 1 Warning
    Quantity 5g
    Concentration Rh 28.2% min.
    Chemical Name or Material Potassium pentachlororhodate(III)
    Assay 99.99%, (metals basis)
    Grade Premion™

    GHS H Statement
    H302-H315-H319
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P301+P312-P321-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Specific treatment (see ... on this label).Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit