Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium pentachlororhodate(III), Premion™, 99.99% (metals basis), Rh 28.2% min

Manufacturer:  Alfa Aesar™ 4040903

Quantity 1g

 View more versions of this product

Catalog No. AA4040903


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

65980-75-8
MFCD00049661
Premion™
358.37
Potassium pentachlororhodate(III)

Specifications

65980-75-8
358.37
Slightly soluble in cold water; Insoluble in alcohol
1g
99.99%, (metals basis)
Powder
MFCD00049661
Rh 28.2% min.
Potassium pentachlororhodate(III)
Premion™

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P301+P312-P321-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Specific treatment (see ... on this label).Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : VI8580332

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit