Alfa Aesar™ Potassium perchlorate, 99%

Manufacturer: Alfa Aesar™ A1129636

Quantity 500g

 View more versions of this product

Catalog No. AAA1129636

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7778-74-7
    Molecular Formula KClO4
    Formula Weight 138.55
    MDL Number MFCD00011362
    UN Number UN1489
    Merck Index 14,7653
    Chemical Name or Material Potassium perchlorate

    Specifications

    Formula Weight 138.55
    Health Hazard 3 GHS P Statement
    P221-P283-P210-P306+P360-P371+P380+P375-P501a
    Take any precaution to avoid mixing with combustibles.
    Wear fire/flame resistant/retardant clothing.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON CLOTHING: rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
    In case of major fire and large quantities: Evacuate area.
    Fight fire remotely due to the risk of explosion.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00011362
    CAS 7778-74-7
    Melting Point 301° to 304°C
    Health Hazard 1 Danger
    RTECS Number SC9700000
    TSCA TSCA
    Odor Odorless
    Molecular Formula KClO4
    UN Number UN1489
    Merck Index 14,7653
    Quantity 500g
    Health Hazard 2 GHS H Statement
    H271-H302
    May cause fire or explosion
    strong oxidizer.
    Harmful if swallowed.
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: POTASSIUM PERCHLORATE
    Chemical Name or Material Potassium perchlorate
    Assay 99%

    Description

    Applications
    Potassium perchlorate is used in ammunition percussion caps, explosive primers, fireworks and propellants. It acts as a solid rocket propellant. It is also used as an oxidizer in colored pyrotechnic compositions and flash powder. It is used as an antithyroid and involved in the treatment of hyperthyroidism. The mixture of anthracene and sulfur with potassium perchlorate is used for generating black smoke signaling.

    Solubility
    Soluble in water. Slightly soluble in acetone. Insoluble in ethanol, ethyl acetate and ether.

    Notes
    Hygroscopic. Incompatible with strong reducing agents, powdered metals, strong acids and alcohols.

    GHS H Statement
    H271-H302
    May cause fire or explosion
    strong oxidizer.
    Harmful if swallowed.

    GHS P Statement
    P221-P283-P210-P306+P360-P371+P380+P375-P501a
    Take any precaution to avoid mixing with combustibles.
    Wear fire/flame resistant/retardant clothing.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    IF ON CLOTHING: rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
    In case of major fire and large quantities: Evacuate area.
    Fight fire remotely due to the risk of explosion.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit