Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium periodate, 99.5%, extra pure, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Plastic bottle
7790-21-8
IKO4
230
FJVZDOGVDJCCCR-UHFFFAOYSA-M
potassium periodate,potassium metaperiodate,unii-q3loc6p66l,q3loc6p66l,periodic acid hio4 , potassium salt,potassium ion io4,ssmicroaea 1/4 o,acmc-20ajue,ssmicroaea 1/4 oni,periodic acid hio4 , potassium salt 1:1
516896
potassium;periodate
[O-]I(=O)(=O)=O.[K+]
7790-21-8
IKO4
230
FJVZDOGVDJCCCR-UHFFFAOYSA-M
potassium periodate,potassium metaperiodate,unii-q3loc6p66l,q3loc6p66l,periodic acid hio4 , potassium salt,potassium ion io4,ssmicroaea 1/4 o,acmc-20ajue,ssmicroaea 1/4 oni,periodic acid hio4 , potassium salt 1:1
516896
potassium;periodate
[O-]I(=O)(=O)=O.[K+]
Oxidation reagent
Assay Percent Range 99.4% min. (Iodometry)
Linear Formula KIO4
Fieser 01,809; 02,311; 04,373; 05,507; 06,455
Merck Index 15,7775
Solubility Information Solubility in water: 0.42 g/100mL (20°C).
Formula Weight 230
Physical Form Crystalline Powder
Percent Purity 99.5%
Grade Extra Pure
Manganese (Mn) 1ppm max.
Other Halogens (as Cl) 0.01% max.
Packaging Plastic bottle
Solubility Solubility in water: 0.42 g/100mL (20°C).
Color White
Melting Point 582.0°C
Quantity 2.5kg
Chemical Name or Material Potassium periodate
Show More Show Less

GHS H Statement
Causes serious eye irritation.
May intensify fire; oxidizer.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occu

GHS Signal Word: Danger

EINECSNumber : 232-196-