Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Permanganate (Crystalline/USP), Fisher Chemical™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Amber Glass
Glass Bottle
7722-64-7
KMnO4
158.032
MFCD00011364
VZJVWSHVAAUDKD-UHFFFAOYSA-N
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
516875
potassium;permanganate
[O-][Mn](=O)(=O)=O.[K+]
7722-64-7
KMnO4
158.032
MFCD00011364
VZJVWSHVAAUDKD-UHFFFAOYSA-N
potassium permanganate, chameleon mineral, condy's crystals, argucide, cairox, permanganate of potash, insta-perm, walko tablets, algae-k, solo san soo
516875
potassium;permanganate
[O-][Mn](=O)(=O)=O.[K+]
Assay Percent Range 99.0 to 100.5 %
Formula Weight 158.03
Physical Form Solid Powder
Percent Purity 99.0 to 100.5%
Grade USP
Identification Pass Test
Insoluble Matter 0.2% max.
Loss on Drying 0.5% max.
Packaging Amber Glass
Residual Solvents Meets Requirements
Color Index No. 77755
Color Brown
Melting Point 240°C
pH 8
Quantity 500g
Chemical Name or Material Potassium Permanganate
Show More Show Less

DANGER!

Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. May cause burns. May be harmful if swallowed. Use only under a chemical fume hood. Wear personal protective equipment. Do not get in eyes, on skin, or on clothing. Avoid ingestion and inhalation. Keep away from clothing and other combustible materials. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:2
Flammability:0
Instability:0

DOTInformation : DOT Class 5.1, : Oxidizer