Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium Permanganate, puriss. p.a. ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 100 g |
UN Number | 1490 |
Formula Weight | 158.03g/mol |
Percent Purity | ≥99% |
Grade | ACS/puriss. p.a. |
Chemical Name or Material | Potassium Permanganate, Reag. Ph. Eur. |
Safety and Handling
P220-P273-P280-P305 + P351 + P338-P310-P501
H272-H302-H314-H410
EINECSNumber : 231-760-3
RTECSNumber : SD6475000