Alfa Aesar™ Potassium peroxydisulfate, 99.99% (metals basis)

Manufacturer: Alfa Aesar™ 1822714

Quantity 25g

 View more versions of this product

Catalog No. AA1822714

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 7727-21-1
    Formula Weight 270.31
    MDL Number MFCD00011386
    UN Number UN1492
    Merck Index 14,7656
    Chemical Name or Material Potassium peroxydisulfate

    Specifications

    Formula Weight 270.31
    Health Hazard 2 GHS H Statement
    H334-H272-H302-H335-H315-H319-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May intensify fire
    oxidizer.
    Harmful if swallowed.
    May cause respiratory irritation.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P221-P210-P284a-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00011386
    Melting Point 100°C (dec.)
    Quantity 25g
    Health Hazard 1 Danger
    Density 2.477g/mL
    TSCA TSCA
    Physical Form Crystalline
    UN Number UN1492
    Merck Index 14,7656
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: POTASSIUM PERSULFATE
    Odor Odorless
    CAS 7727-21-1
    RTECS Number SE0400000
    Chemical Name or Material Potassium peroxydisulfate
    Assay 99.99%, (metals basis)

    GHS H Statement
    H334-H272-H302-H335-H315-H319-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May intensify fire
    oxidizer.
    Harmful if swallowed.
    May cause respiratory irritation.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.

    GHS P Statement
    P221-P210-P284a-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit