Alfa Aesar™ Potassium peroxydisulfate, low nitrogen, ACS, 99.0% min

Manufacturer: Alfa Aesar™ 46939A1

Quantity 1kg

 View more versions of this product

Catalog No. AA46939A1

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7727-21-1
    Molecular Formula K2O8S2
    Linear Formula K2S2O8
    Formula Weight 270.31
    MDL Number MFCD00011386
    UN Number UN1492
    Merck Index 14,7656
    Synonym Potassium persulfate
    Chemical Name or Material Potassium peroxydisulfate, Low nitrogen
    Grade ACS Grade

    Specifications

    Formula Weight 270.31
    Health Hazard 2 GHS H Statement
    H334-H272-H302-H335-H315-H319-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May intensify fire
    oxidizer.
    Harmful if swallowed.
    May cause respiratory irritation.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P221-P210-P284a-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Physical Form Crystalline
    UN Number UN1492
    Density 2.477g/mL
    CAS 7727-21-1
    Merck Index 14,7656
    Odor Odorless
    Health Hazard 1 Danger
    Molecular Formula K2O8S2
    Linear Formula K2S2O8
    MDL Number MFCD00011386
    Synonym Potassium persulfate
    Color White
    Quantity 1kg
    RTECS Number SE0400000
    Melting Point 100°C (dec.)
    DOT Information Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: POTASSIUM PERSULFATE
    Chemical Name or Material Potassium peroxydisulfate, Low nitrogen
    Assay ≥99.0%
    Grade ACS Grade

    GHS H Statement
    H334-H272-H302-H335-H315-H319-H317
    May cause allergy or asthma symptoms or breathing difficulties if inhaled.
    May intensify fire
    oxidizer.
    Harmful if swallowed.
    May cause respiratory irritation.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause an allergic skin reaction.

    GHS P Statement
    P221-P210-P284a-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit