Alfa Aesar™ Potassium perrhenate, 99% (metals basis), Re 64%

A powerful oxidizer

Manufacturer: Alfa Aesar™ 11411SS

Quantity 0.2g

 View more versions of this product

Catalog No. AA11411SS

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10466-65-6
    Formula Weight 289.3
    MDL Number MFCD00011365
    UN Number UN1479
    Chemical Name or Material Potassium perrhenate

    Specifications

    Formula Weight 289.3
    Health Hazard 3 GHS P Statement
    P221-P210-P220-P280-P370+P378a-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H272
    May intensify fire
    oxidizer.
    TSCA TSCA
    Odor Odorless
    MDL Number MFCD00011365
    Melting Point 550°C
    DOT Information Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
    Physical Form Crystalline
    Refractive Index 1.643
    UN Number UN1479
    Boiling Point 1360° to 1370°C
    Health Hazard 1 Danger
    Concentration Re 64%
    Density 4.887g/mL
    CAS 10466-65-6
    Color White
    Quantity 0.2g
    RTECS Number SE1400000
    Chemical Name or Material Potassium perrhenate
    Assay 99%, (metals basis)

    GHS H Statement
    H272
    May intensify fire
    oxidizer.

    GHS P Statement
    P221-P210-P220-P280-P370+P378a-P501a
    Take any precaution to avoid mixing with combustibles.
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Keep/Store away from clothing/combustible materials.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit