Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium perruthenate(VII), 97%, Thermo Scientific Chemicals

Catalog Number AA1187703
Click to view available options
Quantity:
1 g
5 g
10378-50-4
C32H36N2O5
528.65
MFCD00061333
PXMOBROHNZPLTM-UHFFFAOYSA-N
4,7-bis benzyloxy-n,n-diisopropyl-1,3-dioxo-2-isoindolinebutyramide
101141422
4-[1,3-dioxo-4,7-bis(phenylmethoxy)isoindol-2-yl]-N,N-di(propan-2-yl)butanamide
CC(C)N(C(C)C)C(=O)CCCN1C(=O)C2=C(OCC3=CC=CC=C3)C=CC(OCC3=CC=CC=C3)=C2C1=O
10378-50-4
C32H36N2O5
528.65
MFCD00061333
PXMOBROHNZPLTM-UHFFFAOYSA-N
4,7-bis benzyloxy-n,n-diisopropyl-1,3-dioxo-2-isoindolinebutyramide
101141422
4-[1,3-dioxo-4,7-bis(phenylmethoxy)isoindol-2-yl]-N,N-di(propan-2-yl)butanamide
CC(C)N(C(C)C)C(=O)CCCN1C(=O)C2=C(OCC3=CC=CC=C3)C=CC(OCC3=CC=CC=C3)=C2C1=O

Potassium perruthenate(VII) is used in the production of carbon nanotubes for flexible solid-state supercapacitors. It is also used as an oxidizing agent in the synthesis of 5-formylcytosine from 5-hydroxymethylcytosine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 440°C (decomposition)
Color Black
Physical Form Crystalline
Quantity 1 g
Assay Percent Range 97%
Solubility Information Slightly soluble in water.
Formula Weight 204.17
Appearance Black crystals
Chemical Name or Material Potassium perruthenate(VII)
Hazard Category H272-H315-H319-H335
Hazard Statement Keep away from combustible material. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P370+P380+P375-P372-P380-P501c
DOTInformation DOT Class: 5.1, Packaging Group: II
EINECSNumber 233-835-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit