Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium persulfate, 99+%, for analysis, ACROS Organics™

Click to view available options
Quantity:
25g
250g
500g
1kg
5kg
Packaging:
Plastic bottle
7727-21-1
K2O8S2
270.3
MFCD00011386
USHAGKDGDHPEEY-UHFFFAOYSA-L
potassium persulfate,anthion,potassium peroxydisulfate,potassium peroxodisulfate,dipotassium peroxydisulfate,potassium peroxydisulphate,dipotassium persulfate,caswell no. 700,dipotassium peroxodisulphate,unii-6b86k0mczc
24412
dipotassium;sulfonatooxy sulfate
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+]
7727-21-1
K2O8S2
270.3
MFCD00011386
USHAGKDGDHPEEY-UHFFFAOYSA-L
potassium persulfate,anthion,potassium peroxydisulfate,potassium peroxodisulfate,dipotassium peroxydisulfate,potassium peroxydisulphate,dipotassium persulfate,caswell no. 700,dipotassium peroxodisulphate,unii-6b86k0mczc
24412
dipotassium;sulfonatooxy sulfate
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+]
Oxidation reagent
Assay Percent Range 99% min. (Iodometry)
Linear Formula K2S2O8
Merck Index 15,7777
Solubility Information Solubility in water: 5g/100mL (20°C). Other solubilities: insoluble in alcohol
Formula Weight 270.3
Physical Form Crystalline Powder
Percent Purity 99+%
Grade Analysis
Chloride 120ppm max
Chlorine (Cl) 50ppm max.
Density 2.47
Heavy Metals (as Pb) 50ppm max.
Iron (Fe) 20ppm max.
Manganese (Mn) 5ppm max.
Packaging Plastic bottle
Solubility Solubility in water: 5g/100mL (20°C). Other solubilities: insoluble in alcohol
Color White
Melting Point 100.0°C
Quantity 250g
Chemical Name or Material Potassium persulfate
Show More Show Less

GHS H Statement
May intensify fire; oxidizer.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficu

GHS P Statement
Keep/Store away from clothing/ combustible materials.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,r

GHS Signal Word: Danger

EINECSNumber : 231-781-8