Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Persulfate, BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly Bottle
Wide Mouth Amber Glass Bottle
7727-21-1
K2O8S2
270.309
USHAGKDGDHPEEY-UHFFFAOYSA-L
potassium persulfate,anthion,potassium peroxydisulfate,potassium peroxodisulfate,dipotassium peroxydisulfate,potassium peroxydisulphate,dipotassium persulfate,caswell no. 700,dipotassium peroxodisulphate,unii-6b86k0mczc
24412
dipotassium;sulfonatooxy sulfate
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+]
7727-21-1
K2O8S2
270.309
USHAGKDGDHPEEY-UHFFFAOYSA-L
potassium persulfate,anthion,potassium peroxydisulfate,potassium peroxodisulfate,dipotassium peroxydisulfate,potassium peroxydisulphate,dipotassium persulfate,caswell no. 700,dipotassium peroxodisulphate,unii-6b86k0mczc
24412
dipotassium;sulfonatooxy sulfate
[O-]S(=O)(=O)OOS(=O)(=O)[O-].[K+].[K+]

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

    Suitable for Phosphate and Mercury Determinations
Formula Weight 270.32
Grade BAKER INSTRA-ANALYZED™ Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.48kg
Quantity 500g
Chemical Name or Material Potassium Persulfate