Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Potassium phosphate, 0.2M buffer soln., pH 7.4, low endotoxin

Click to view available options
Quantity:
100mL
250mL
7778-77-0
H2KO4P
136.084
MFCD00011401
GNSKLFRGEWLPPA-UHFFFAOYSA-M
516951
CHEBI:63036
potassium;dihydrogen phosphate
OP(=O)(O)[O-].[K+]
7778-77-0
H2KO4P
136.084
MFCD00011401
GNSKLFRGEWLPPA-UHFFFAOYSA-M
516951
CHEBI:63036
potassium;dihydrogen phosphate
OP(=O)(O)[O-].[K+]
Concentration 0.2M buffer soln.
Physical Form Liquid
pH 7.4
Color Colorless
Quantity 100mL
Chemical Name or Material Potassium phosphate, low endotoxin