Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium phosphate, dibasic, 99+%, for analysis, anhydrous, ACROS Organics™
Click to view available options
Quantity:
250g
1kg
5kg
Packaging:
Plastic bottle
Plastic drum
Description
Potassium phosphate, dibasic, 99+%, for analysis, anhydrous, ACROS Organics, Quantity: 5kg, Packaging: Plastic drum, CAS: 7758-11-4, White, Grade: Analysis, Melting Point: 340.0 deg.C, Molecular Formula: HK2O4P, Molecular Weight: 174.18, Percent Purity: 99+%, pH: 8.5 to 9.6 (5% soln. at 20 deg.C)
For analysisSpecifications
Assay Percent Range | 99+% |
Linear Formula | K2HPO4 |
Merck Index | 15,7779 |
Solubility Information | Solubility in water: 1600g/L (20°C). Other solubilities: slightly soluble in alcohol |
Formula Weight | 174.18 |
Physical Form | Fine Crystalline Powder |
Percent Purity | 99+% |
Grade | Analysis |
Chloride | 50ppm max |
Chlorine (Cl) | 50ppm max. |
Show More |
Safety and Handling
EINECSNumber : 231-834-5