Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate, Dibasic, Endotoxin Tested, Multi-Compendial, U.S.P., J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
5kg
Packaging:
Poly Bottle
Poly Pail
7758-11-4
HK2O4P
174.17
ZPWVASYFFYYZEW-UHFFFAOYSA-L
dipotassium hydrogen phosphate,dipotassium phosphate,dipotassium hydrogenphosphate,dibasic potassium phosphate,potassium hydrogen phosphate,potassium phosphate, dibasic,potassium dibasic phosphate,potassium phosphate dibasic,phosphoric acid, dipotassium salt,dipotassium monophosphate
24450
CHEBI:32031
dipotassium;hydrogen phosphate
OP(=O)([O-])[O-].[K+].[K+]
7758-11-4
HK2O4P
174.17
ZPWVASYFFYYZEW-UHFFFAOYSA-L
dipotassium hydrogen phosphate,dipotassium phosphate,dipotassium hydrogenphosphate,dibasic potassium phosphate,potassium hydrogen phosphate,potassium phosphate, dibasic,potassium dibasic phosphate,potassium phosphate dibasic,phosphoric acid, dipotassium salt,dipotassium monophosphate
24450
CHEBI:32031
dipotassium;hydrogen phosphate
OP(=O)([O-])[O-].[K+].[K+]
Formula Weight 174.18
Grade Multi-Compendial/USP
Packaging Poly Bottle
Quantity 2.5kg
Chemical Name or Material Potassium Phosphate, Dibasic, Endotoxin Tested