Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate, Dibasic, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
2.5kg
500g
Packaging:
Poly Bottle

Chemical Identifiers

7758-11-4
HK2O4P
174.17
ZPWVASYFFYYZEW-UHFFFAOYSA-L
24450
CHEBI:32031

High quality chemicals for laboratory and specialized industrial use

CAS 7758-11-4
Molecular Formula HK2O4P
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
SMILES OP(=O)([O-])[O-].[K+].[K+]
IUPAC Name dipotassium;hydrogen phosphate
Molecular Weight (g/mol) 174.17
PubChem CID 24450
ChEBI CHEBI:32031
Formula Weight 174.18
Physical Form Powder
Packaging Poly Bottle
Specific Gravity 1L = 1.00kg
Quantity 2.5kg
Chemical Name or Material Potassium Phosphate, Dibasic
Grade Baker Analyzed™ ACS Reagent
Show More Show Less