Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate Dibasic, Purified (Dipotassium Hydrogen Phosphate), Reagents

Catalog No. p-200021062
Click to view available options
Quantity:
100 g
500 g

Potassium Phosphate Dibasic, Purified (Dipotassium Hydrogen Phosphate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7758-11-4
Molecular Formula HK2O4P
Molecular Weight (g/mol) 174.17
InChI Key ZPWVASYFFYYZEW-UHFFFAOYSA-L
Synonym Dipotassium Hydrogen Phosphate
IUPAC Name dipotassium hydrogen phosphate
SMILES [K+].[K+].OP([O-])([O-])=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 100 g
Grade Pure
Packaging Glass Bottle
Specific Gravity 2.3
Chemical Name or Material Potassium Phosphate Dibasic, Purified (Dipotassium Hydrogen Phosphate)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.