Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate Dibasic, Trihydrate, Reagent, 98%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
16788-57-1
H7K2O7P
228.22
XQGPKZUNMMFTAL-UHFFFAOYSA-L
dipotassium trihydrate hydrogen phosphate
O.O.O.[K+].[K+].OP([O-])([O-])=O
16788-57-1
H7K2O7P
228.22
XQGPKZUNMMFTAL-UHFFFAOYSA-L
dipotassium trihydrate hydrogen phosphate
O.O.O.[K+].[K+].OP([O-])([O-])=O

P1385, 16788-57-1, K2HPO4•3H2O

Spectrum™ Potassium Phosphate Dibasic, Trihydrate, Reagent is a soluble salt that is commonly used as a buffering agent. As a Reagent Grade, Spectrum Chemical manufactured potassium phosphate is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Grade Reagent
CAS Max % 1
Percent Purity 98%
Heavy Metals (as Pb) 5e-006
Insoluble Matter 0.0001
Iron (Fe) 1e-005
Nitrogen (N) 1e-005
Packaging Poly Pail
Sodium (Na) 0.0005
Sulfate 5e-005
pH 9.0 to 9.4
Quantity 12 kg
Show More Show Less