Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate Monobasic, ACS Reagent (Potassium Dihydrogen Phosphate), Reagents

Catalog No. p-200021020
Click to view available options
Quantity:
100 g
500 g
3 kg
10 kg
Packaging:
Glass Bottle
Poly Bottle
Poly Pail

Potassium Phosphate Monobasic, ACS Reagent (Potassium Dihydrogen Phosphate), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Potassium Phosphate Monobasic, ACS Reagent (Potassium Dihydrogen Phosphate)
Boiling Point 400°C (loses water and becomes metaphosphate)
Melting Point 253°C
Color Colorless/White
CAS 7778-77-0
pH 4.1 to 4.5
Physical Form Solid
Quantity 100 g
Molecular Formula KH2PO4
Synonym Potassium Dihydrogen Phosphate
Molecular Weight (g/mol) 136.0855
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.338
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.