Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate, Monobasic, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500g
2.5kg
Packaging:
Poly bottle
7778-77-0
H2KO4P
136.084
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate
516951
CHEBI:63036
potassium;dihydrogen phosphate
OP(=O)(O)[O-].[K+]
7778-77-0
H2KO4P
136.084
GNSKLFRGEWLPPA-UHFFFAOYSA-M
potassium dihydrogen phosphate,monopotassium phosphate,potassium phosphate monobasic,potassium phosphate, monobasic,potassium acid phosphate,phosphoric acid, monopotassium salt,monobasic potassium phosphate,monopotassium monophosphate,monopotassium dihydrogen phosphate,monopotassium orthophosphate
516951
CHEBI:63036
potassium;dihydrogen phosphate
OP(=O)(O)[O-].[K+]

High quality chemicals for laboratory and specialized industrial use

Formula Weight 136.09
Physical Form Crystals
Grade Meets ACS reagent chemical requirements
Packaging Poly bottle
Specific Gravity 1L = 2.34kg
Quantity 2.5kg
Chemical Name or Material Potassium Phosphate, Monobasic