Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium Phosphate, Monobasic, ULTREX™ Ultrapure Reagent, J.T.Baker™

Click to view available options
Quantity:
100g
1kg
Packaging:
Wide Mouth Amber Glass Bottle
7778-77-0
H2KO4P
136.084
GNSKLFRGEWLPPA-UHFFFAOYSA-M
516951
CHEBI:63036
potassium;dihydrogen phosphate
OP(=O)(O)[O-].[K+]
7778-77-0
H2KO4P
136.084
GNSKLFRGEWLPPA-UHFFFAOYSA-M
516951
CHEBI:63036
potassium;dihydrogen phosphate
OP(=O)(O)[O-].[K+]
Formula Weight 136.09
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.34kg
Quantity 100g
Grade ULTREX™ Ultrapure Reagent
Chemical Name or Material Potassium Phosphate, Monobasic